Skip to Content
Merck
All Photos(1)

Key Documents

A0363700

Ambroxol hydrochloride

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

2-Amino-3,5-dibromo-N-(trans-4-hydroxycyclohexyl)benzylamine

Sign Into View Organizational & Contract Pricing

Select a Size

125 MG
$254.00

$254.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
125 MG
$254.00

About This Item

Empirical Formula (Hill Notation):
C13H18Br2N2O · HCl
CAS Number:
Molecular Weight:
414.56
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$254.00


Check Cart for Availability

Request a Bulk Order

grade

pharmaceutical primary standard

API family

ambroxol

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Cl.Nc1c(Br)cc(Br)cc1CN[C@@H]2CC[C@@H](O)CC2

InChI

1S/C13H18Br2N2O.ClH/c14-9-5-8(13(16)12(15)6-9)7-17-10-1-3-11(18)4-2-10;/h5-6,10-11,17-18H,1-4,7,16H2;1H/t10-,11-;

InChI key

QNVKOSLOVOTXKF-PFWPSKEQSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Ambroxol hydrochloride EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jan Lukas et al.
Molecular therapy : the journal of the American Society of Gene Therapy, 23(3), 456-464 (2014-11-21)
Lysosomal storage disorders (LSD) are a group of heterogeneous diseases caused by compromised enzyme function leading to multiple organ failure. Therapeutic approaches involve enzyme replacement (ERT), which is effective for a substantial fraction of patients. However, there are still concerns
Julien Arnold et al.
Artificial cells, nanomedicine, and biotechnology, 43(1), 26-32 (2014-08-01)
A gastro-retentive capsule has been prepared which is retained in the stomach for a period of 24h, providing a vehicle for the controlled delivery to the upper intestines. These "gastro cocoons" can resist passage through the sphincter of the stomach

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service