Key Documents
99908
Supelco
Developing tank of glass with lid for TLC
About This Item
UNSPSC Code:
41115711
size
17.5 cm × 6.2 cm × 6.8 cm
General description
TLC developing tanks are used to run TLC plates for separation. It is important to maintain saturated conditions during the analysis which can be achieved by using small tanks, filter paper liners and sufficient solvents. The tank can be equilibrated for 30mins prior to running the plate to yield good results.[1] Lid is used to cover the tank.
Application
Developing tank of glass with lid for TLC may be used to develop TLC plates.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
An overview on thin layer chromatography
Bele, Archana A., and Anubha Khale.
International Journal of Pharmaceutical Sciences and Research, 2, 256-267 (2011)