Skip to Content
Merck
All Photos(1)

Key Documents

98837

Supelco

4′-Hydroxyacetophenone

analytical standard

Synonym(s):

4-Hydroxyphenylethanone, p-Acetophenol, p-Hydroxyphenyl methyl ketone, Piceol

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$98.40

$98.40


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
$98.40

About This Item

Linear Formula:
HOC6H4COCH3
CAS Number:
Molecular Weight:
136.15
Beilstein:
774355
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

$98.40


Check Cart for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥98.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

147-148 °C/3 mmHg (lit.)

mp

107-110 °C
109-111 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

CC(=O)c1ccc(O)cc1

InChI

1S/C8H8O2/c1-6(9)7-2-4-8(10)5-3-7/h2-5,10H,1H3

InChI key

TXFPEBPIARQUIG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

330.8 °F

Flash Point(C)

166 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Dao Feng Xiang et al.
Biochemistry, 53(21), 3476-3485 (2014-05-17)
The uncharacterized protein Rsp3690 from Rhodobacter sphaeroides is a member of the amidohydrolase superfamily of enzymes. In this investigation the gene for Rsp3690 was expressed in Escherichia coli and purified to homogeneity, and the three-dimensional structure was determined to a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service