Skip to Content
Merck
All Photos(1)

Key Documents

95293

Supelco

Hydrogen ionophore I - cocktail B

Selectophore

Synonym(s):

H⁺ ionophore I - cocktail B

Sign Into View Organizational & Contract Pricing

Select a Size

0.1 ML
$474.00

$474.00


Available to ship on04 April 2025Details


Request a Bulk Order

Select a Size

Change View
0.1 ML
$474.00

About This Item

UNSPSC Code:
26111700
NACRES:
NB.61

$474.00


Available to ship on04 April 2025Details


Request a Bulk Order

product line

Selectophore

Quality Level

form

liquid

composition

Hydrogen ionophore I, 10.0 wt. % (95292)
2-Nitrophenyl octyl ether, 89.3 wt. % (73732)
Potassium tetrakis(4-chlorophenyl)borate, 0.7 wt. % (60591)

General description

Visit our Sensor Applications portal to learn more.

Application

Selected application examples

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gang Sa et al.
The New phytologist, 222(4), 1951-1964 (2019-02-14)
Salt stress is an important environmental cue impeding poplar nitrogen nutrition. Here, we characterized the impact of salinity on proton-driven nitrate fluxes in ectomycorrhizal roots and the importance of a Hartig net for nitrate uptake. We employed two Paxillus involutus
A hydrogen ion-selective liquid-membrane electrode based on tri-n-dodecylamine as neutral carrier
Schulthess, P.; et al.
Analytica Chimica Acta, 131, 111-116 (1981)
Neutral carrier based hydrogen ion selective microelectrode for extra- and intracellular studies.
D Ammann et al.
Analytical chemistry, 53(14), 2267-2269 (1981-12-01)
Robert D Hoffmann et al.
Nature communications, 11(1), 2395-2395 (2020-05-16)
Pollen tubes are highly polarized tip-growing cells that depend on cytosolic pH gradients for signaling and growth. Autoinhibited plasma membrane proton (H+) ATPases (AHAs) have been proposed to energize pollen tube growth and underlie cell polarity, however, mechanistic evidence for

Questions

1–2 of 2 Questions  
  1. What are the formulations for Products 95291, Hydrogen ionophore I - cocktail A and 95293, Hydrogen ionophore I - cocktail B, and how do they differ?

    1 answer
    1. Hydrogen ionophore I cocktails A and B are quite similar. Product 95291 is Ionophore I (tridodecylamine, Product 95292), 10%; 2-Nitrophenyl-octylether (Product 73732), 89.3%, and sodium tetraphenylborate (Product 72018), 0.7%. For Product 95293, the first two components are the same; the third component is potassium tetrakis(4-chlorophenyl)borate, 0.7%.

      Helpful?

  2. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service