Skip to Content
Merck
All Photos(1)

Documents

95038

Sigma-Aldrich

Propyl 4-hydroxybenzoate sodium salt

tested according to Ph. Eur.

Synonym(s):

Propylis parahydroxybenzoas natricum, Sodium propyl 4-hydroxybenzoate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H11NaO3
CAS Number:
Molecular Weight:
202.18
Beilstein:
5183350
EC Number:
MDL number:
UNSPSC Code:
12352107
PubChem Substance ID:
NACRES:
NA.21

Agency

USP/NF
tested according to Ph. Eur.

Quality Level

form

crystals

application(s)

pharmaceutical (small molecule)

SMILES string

[Na+].CCCOC(=O)c1ccc([O-])cc1

InChI

1S/C10H12O3.Na/c1-2-7-13-10(12)8-3-5-9(11)6-4-8;/h3-6,11H,2,7H2,1H3;/q;+1/p-1

InChI key

IXMINYBUNCWGER-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anna Maria Wróbel et al.
Toxicology letters, 230(3), 375-381 (2014-08-17)
Numerous studies have shown that widely used parabens possess estrogenic properties. In the present study, we examined the effects of methyl-, propyl- and butylparaben on the mRNA and protein expression of estrogen receptor (ER)-α (ESR1) and -β (ESR2) and the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service