Skip to Content
Merck
All Photos(1)

Key Documents

94348

Supelco

N-(3-Phenylpropionyl)glycine

analytical standard

Synonym(s):

2-[(3-Phenylpropionyl)amino]acetic acid, 3-Phenylpropionyl glycine, N-(1-Oxo-3-phenylpropyl)glycine, N-(3-Phenylpropanoyl)glycine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H13NO3
CAS Number:
Molecular Weight:
207.23
Beilstein:
2976435
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:

grade

analytical standard

Quality Level

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

application(s)

clinical testing

format

neat

storage temp.

2-8°C

InChI

1S/C11H13NO3/c13-10(12-8-11(14)15)7-6-9-4-2-1-3-5-9/h1-5H,6-8H2,(H,12,13)(H,14,15)

InChI key

YEIQSAXUPKPPBN-UHFFFAOYSA-N

Biochem/physiol Actions

Phenylpropionylglycine is an acylglycine. Acylglycines are normally minor metabolites of fatty acids. However, the excretion of certain acylglycines is increased in several inborn errors of metabolism. In certain cases the measurement of these metabolites in body fluids can be used to diagnose disorders associated with mitochondrial fatty acid beta-oxidation. Acyl glycines are produced through the action of glycine N-acyltransferase (EC 2.3.1.13) which is an enzyme that catalyzes the chemical reaction: acyl-CoA + glycine ↔ CoA + N-acylglycine. The detection of phenylpropionylglycine in urine after an oral load of phenylpropionic acid can be used to diagnose deficiency of medium-chain acyl-CoA dehydrogenase, a frequent and treatable metabolic defect.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B Flath et al.
Journal of chromatography. B, Biomedical sciences and applications, 694(1), 227-232 (1997-06-20)
Deficiency of medium-chain acyl-CoA dehydrogenase is a frequent and treatable metabolic defect, which can be diagnosed by detection of phenylpropionylglycine in urine after an oral load of phenylpropionic acid. We studied the determination of phenylpropionylglycine in urine by isocratic ion-exclusion

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service