Skip to Content
Merck
All Photos(1)

Documents

94113

Supelco

Eriocitrin

analytical standard

Synonym(s):

(S)-3′,4′,5,7-Tetrahydroxyflavanone-7-[6-O-(α-L-rhamnopyranosyl)-β-D-glucopyranoside], Eriodictioside, Eriodictyol 7-O-rutinoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H32O15
CAS Number:
Molecular Weight:
596.53
Beilstein:
1304401
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤10.0% water

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

C[C@@H]1O[C@@H](OC[C@H]2O[C@@H](Oc3cc(O)c4C(=O)C[C@H](Oc4c3)c5ccc(O)c(O)c5)[C@H](O)[C@@H](O)[C@@H]2O)[C@H](O)[C@H](O)[C@H]1O

InChI

1S/C27H32O15/c1-9-20(32)22(34)24(36)26(39-9)38-8-18-21(33)23(35)25(37)27(42-18)40-11-5-14(30)19-15(31)7-16(41-17(19)6-11)10-2-3-12(28)13(29)4-10/h2-6,9,16,18,20-30,32-37H,7-8H2,1H3/t9-,16-,18+,20-,21+,22+,23-,24+,25+,26+,27+/m0/s1

InChI key

OMQADRGFMLGFJF-MNPJBKLOSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Eriocitrin is a flavonoid glycoside mainly found in fruits like lemon.

Application

Eriocitrin may be used as a reference standard in the determination of eriocitrin in samples of lime juice using solid phase extraction coupled with high performance liquid chromatography (SPE-HPLC).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Flavone glycoside found in lemon. Administration prior to acute exercise helped to maintain the redox status of liver glutathione. Without eriocitrin, it shifts markedly toward oxidized form.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Identification and antioxidant activity of flavonoid metabolites in plasma and urine of eriocitrin-treated rats.
Miyake Y, et al.
Journal of Agricultural and Food Chemistry, 48(8), 3217-3224 (2000)
Iman Saeidi et al.
Journal of pharmaceutical and biomedical analysis, 56(2), 419-422 (2011-06-21)
Solid phase extraction (SPE) and HPLC were used for simultaneous determination of hesperidin, diosmin and eriocitrin in Iranian lime juice samples. The method involved very simple efficient SPE with polyamide cartridge, the use of mixture of water/acetonitrile/acetic acid (78:19:3, v/v)
Su-Chen Ho et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 71, 176-182 (2014-06-24)
Inhibiting microglial activation-mediated neuroinflammation has become a convincing target for the development of functional foods to treat neurodegenerative diseases. Tangerine peel (Citri reticulatae pericarpium) has potent anti-inflammatory capacity; however, its anti-neuroinflammatory capacity and the corresponding active compounds remain unclear. To
Marely G Figueroa-Pérez et al.
Journal of agricultural and food chemistry, 62(49), 12027-12033 (2014-12-03)
Peppermint (Mentha piperita) infusions represent an important source of antioxidants, which can be enhanced by inducing abiotic stress in plants. The aim of this study was to evaluate the effect of drought stress on peppermint cultivation as well as the
Wanpeng Xi et al.
Food chemistry, 161, 230-238 (2014-05-20)
China is one of the most important diversity centres of the genus Citrus L. and is particularly rich in pummelo germplasm. In this study, the flavonoids in the peels and pulps of 28 Chinese local pummelos and four grapefruits were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service