93657
Tryptone Medium
suitable for microbiology, NutriSelect® Basic
About This Item
Recommended Products
sterility
non-sterile
Quality Level
product line
BioChemika
form
powder
shelf life
limited shelf life, expiry date on the label
composition
sodium chloride, 8 g/L
tryptone, 10 g/L
manufacturer/tradename
NutriSelect® Basic
technique(s)
microbe id | specific enzyme detection: suitable
microbiological culture: suitable
application(s)
agriculture
environmental
food and beverages
water monitoring
microbiology
suitability
nonselective for Enterobacter spp.
nonselective for Escherichia coli
nonselective for Proteus spp.
nonselective for bacteria (General Media)
nonselective for coliforms
nonselective for enterobacteriaceae
Application
Preparation Note
Footnote
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
An article concerning selective growth media for differentiation and detection of Escherichia coli and other coliforms.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service