Skip to Content
Merck
All Photos(1)

Key Documents

93555

Sigma-Aldrich

Tropolone

purum, ≥98.0% (GC)

Synonym(s):

2-Hydroxy-2,4,6-cycloheptatrien-1-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H6O2
CAS Number:
Molecular Weight:
122.12
Beilstein:
1904978
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

purum

Quality Level

Assay

≥98.0% (GC)

form

crystals

bp

80-84 °C/0.1 mmHg (lit.)

mp

50-52 °C (lit.)
50-52 °C

storage temp.

2-8°C

SMILES string

OC1=CC=CC=CC1=O

InChI

1S/C7H6O2/c8-6-4-2-1-3-5-7(6)9/h1-5H,(H,8,9)

InChI key

MDYOLVRUBBJPFM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

233.6 °F - closed cup

Flash Point(C)

112 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Tomasz Borowski et al.
Journal of biological inorganic chemistry : JBIC : a publication of the Society of Biological Inorganic Chemistry, 17(6), 881-890 (2012-05-25)
6-Hydroxymethyl-6-methylcyclohexa-2,4-dienone is a mechanistic probe which when incubated with an extradiol dioxygenase yields a 2-tropolone product. This observation was originally interpreted as evidence supporting a direct heterolytic 1,2-alkenyl migration mechanism for a ring expansion reaction catalyzed by this class of
Eric Sabondjian et al.
Contrast media & molecular imaging, 7(1), 76-84 (2012-02-22)
A challenge with cardiac cell therapy is determining the location of cells relative to infarct tissue. As cells are viable following ¹¹¹In-labeling, and first-pass CT imaging can identify regions of myocardial infarction, we evaluated the feasibility of a SPECT/CT system
Jack Davison et al.
Proceedings of the National Academy of Sciences of the United States of America, 109(20), 7642-7647 (2012-04-18)
A gene cluster encoding the biosynthesis of the fungal tropolone stipitatic acid was discovered in Talaromyces stipitatus (Penicillium stipitatum) and investigated by targeted gene knockout. A minimum of three genes are required to form the tropolone nucleus: tropA encodes a
Christine Meck et al.
Organic letters, 14(23), 5988-5991 (2012-11-22)
α-Hydroxytropolones are a class of molecules with therapeutic potential against several human diseases. However, structure-activity relationship studies on these molecules have been limited due to a scarcity of efficient synthetic methods to access them. It is demonstrated herein that α-hydroxytropolones
Mohammad R Seyedsayamdost et al.
Journal of the American Chemical Society, 133(45), 18343-18349 (2011-09-21)
Marine bacteria and microalgae engage in dynamic symbioses mediated by small molecules. A recent study of Phaeobacter gallaeciensis, a member of the large roseobacter clade of α-proteobacteria, and Emiliania huxleyi, a prominent member of the microphytoplankton found in large algal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service