Skip to Content
Merck
All Photos(1)

Key Documents

93103

Supelco

Paraoxon-ethyl-d10 (diethyl-d10)

PESTANAL®, analytical standard

Synonym(s):

O,O-Diethyl-d10 O-(4-nitrophenyl) phosphate, Diethyl-d10 p-nitrophenyl phosphate, Ethyl-d10 paraoxon

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$397.00

$397.00


0 left in stock

Request a Bulk Order

Select a Size

Change View
5 MG
$397.00

About This Item

Empirical Formula (Hill Notation):
C10D10H4NO6P
Molecular Weight:
285.26
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$397.00


0 left in stock

Request a Bulk Order

grade

analytical standard

Quality Level

product line

PESTANAL®

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

application(s)

agriculture

format

neat

storage temp.

2-8°C

General description

Paraoxon-ethyl-d10 (diethyl-d10) is an isotope-labeled analog of organophosphate pesticide paraoxon, wherein the diethoxy protons are replaced by deuterium.[1][2]

Application

Paraoxon-ethyl-d10 (diethyl-d10) may be used as a deuterated internal standard to quantify the paraoxon-ethyl in animal biological samples by liquid chromatography-high resolution mass spectrometry (LC-HRMS) technique.[3]
Isotope-labeled compounds are increasingly used in isotope dilution mass spectrometry (IDMS) for the quantitative analysis of pesticides. The major advantage of using this technique is that the isotope-labeled compounds have nearly the same physical properties as their non-labeled counterpart analogs, and thus show identical behavior during the workup and sample preparation process. This helps in overcoming the problems of matrix effects generally encountered in the usual LC-MS/GC-MS analysis potentially resulting in biased results. By spiking the sample before workup with its isotope labeled analog, the loss of analyte that leads to matrix effects can be determined and compensated.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 1 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enzyme-free electrochemical detection of nanomolar levels of the organophosphorus pesticide paraoxon-ethyl by using a poly (N-isopropyl acrylamide)-chitosan microgel decorated with palladium nanoparticles
Mutharani B, et al.
Microchimica Acta, 186(3), 1-11 (2019)
Interaction of Cucurbit [7] uril with Oxime K027, Atropine, and Paraoxon: Risky or Advantageous Delivery System?
Zdarova K, et al.
International Journal of Molecular Sciences, 21(21), 7883-7883 (2020)
4-Hydroxyestrone, an Endogenous Estrogen Metabolite, Can Strongly Protect Neuronal Cells Against Oxidative Damage
Choi HJ, et al.
Scientific reports, 10(1), 1-15 (2020)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service