Skip to Content
Merck
All Photos(1)

Key Documents

91977

Supelco

Perfluorononanoic acid

analytical standard

Synonym(s):

Heptadecafluorononanoic acid, Perfluorononanoic acid

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
$169.00

$169.00


0 left in stock

Request a Bulk Order

Select a Size

Change View
50 MG
$169.00

About This Item

Linear Formula:
CF3(CF2)7COOH
CAS Number:
Molecular Weight:
464.08
Beilstein:
1897287
EC Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

$169.00


0 left in stock

Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥96.5% (GC)
96.5-103.0% (T)

shelf life

limited shelf life, expiry date on the label

mp

59-62 °C (lit.)

application(s)

PFAS testing

format

neat

SMILES string

OC(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

InChI

1S/C9HF17O2/c10-2(11,1(27)28)3(12,13)4(14,15)5(16,17)6(18,19)7(20,21)8(22,23)9(24,25)26/h(H,27,28)

InChI key

UZUFPBIDKMEQEQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - Lact. - Repr. 1B - STOT RE 1

Target Organs

Liver

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Liquid Chromatography–Mass Spectrometry (LC-MS) separation of 33 PFAS (Per- and poly-fluoroalkyl substances) using a superficially porous silica particles (SPP) based Ascentis® Express C18 column.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service