Skip to Content
Merck
All Photos(1)

Key Documents

91842

Supelco

Naringin

analytical standard

Synonym(s):

4′,5,7-Trihydroxyflavanone 7-rhamnoglucoside, Naringenin 7-neohesperidoside, Naringenine-7-rhamnosidoglucoside, Naringoside

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
$169.00

$169.00


Available to ship on07 April 2025Details


Request a Bulk Order

Select a Size

Change View
500 MG
$169.00

About This Item

Empirical Formula (Hill Notation):
C27H32O14
CAS Number:
Molecular Weight:
580.53
Beilstein:
102012
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

$169.00


Available to ship on07 April 2025Details


Request a Bulk Order

Quality Level

grade

analytical standard

Assay

≥95.0% (HPLC)

optical activity

[α]/D -80±10°, c = 1 in ethanol

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

C[C@@H]1O[C@@H](O[C@@H]2[C@@H](O)[C@H](O)[C@@H](CO)O[C@H]2Oc3cc(O)c4C(=O)CC(Oc4c3)c5ccc(O)cc5)[C@H](O)[C@H](O)[C@H]1O

InChI

1S/C27H32O14/c1-10-20(32)22(34)24(36)26(37-10)41-25-23(35)21(33)18(9-28)40-27(25)38-13-6-14(30)19-15(31)8-16(39-17(19)7-13)11-2-4-12(29)5-3-11/h2-7,10,16,18,20-30,32-36H,8-9H2,1H3/t10-,16?,18+,20-,21+,22+,23-,24+,25+,26-,27+/m0/s1

InChI key

DFPMSGMNTNDNHN-JJLSSNRUSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Naringin is a naturally occurring flavone glycoside[1][2], which may possess many pharmacological properties like anti-inflammatory, anti-oxidant and anti-apoptotic activity.[2] It is widely found in fruits like grapes and oranges.[2]

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Naringin, a flavanoid in grapefruit and other citrus fruits, potently inhibits intestinal organic anion-transporting polypeptide 1A2 (OATP1A2). Grapefruit juice thereby reduces bioavailability of many pharmacological agents taken at the same time.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Naringin ameliorates diabetic nephropathy by inhibiting NADPH oxidase 4
Zhang J, et al.
European Journal of Pharmacology, 804, 1-6 (2017)
Therapeutic effects of silymarin and naringin on methotrexate?induced nephrotoxicity in rats: Biochemical evaluation of anti?inflammatory, antiapoptotic, and antiautophagic properties
Kandemir MF, et al.
Journal of Food Biochemistry, 41 (2017)
Yi-Chu Nie et al.
Journal of medicinal food, 15(10), 894-900 (2012-09-19)
Naringin, a well-known flavanone glycoside of grapefruit and citrus fruits, was found to be as an effective anti-inflammatory compound in our previous lipopolysaccharide-induced acute lung injury mouse model via blockading activity of nuclear factor κB. The current study sought to
Ayman M Mahmoud et al.
Journal of diabetes and its complications, 26(6), 483-490 (2012-07-20)
Abnormal regulation of glucose and impaired carbohydrate utilization that result from a defective or deficient insulin are the key pathogenic events in type 2 diabetes mellitus (T2DM). The present study was hypothesized to investigate the beneficial effects of hesperidin and
Jiukai Zhang et al.
Food chemistry, 135(3), 1471-1478 (2012-09-08)
Huyou (Citrus changshanensis) is rich in naringin and neohesperidin, which are natural flavanone glycosides with a range of biological activities. Among the different fruit parts, i.e. flavedo, albedo, segment membrane (SM), and juice sacs (JS), albedo showed the highest contents

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service