Skip to Content
Merck
All Photos(1)

Key Documents

91746

Sigma-Aldrich

Triisopropylsilyl trifluoromethanesulfonate

purum, ≥98.0% (T)

Synonym(s):

TIPS triflate, Trifluoromethanesulfonic acid triisopropylsilyl ester, Triisopropylsilyl triflate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3SO3Si[CH(CH3)2]3
CAS Number:
Molecular Weight:
306.42
Beilstein:
3591541
MDL number:
UNSPSC Code:
12352000

grade

purum

Assay

≥98.0% (T)

refractive index

n20/D 1.415 (lit.)

bp

45-46 °C/0.03 mmHg

density

1.14 g/mL at 25 °C (lit.)

SMILES string

CC(C)[Si](OS(=O)(=O)C(F)(F)F)(C(C)C)C(C)C

InChI

1S/C10H21F3O3SSi/c1-7(2)18(8(3)4,9(5)6)16-17(14,15)10(11,12)13/h7-9H,1-6H3

InChI key

LHJCZOXMCGQVDQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Reagent for the smooth introduction of the TIPS group; Review

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R. Bruckmann et al.
Chemische Berichte, 120, 635-635 (1987)
J.M. Muchowski et al.
Tetrahedron Letters, 26, 5375-5375 (1985)
E.J. Corey et al.
Tetrahedron Letters, 22, 3455-3455 (1981)
G. Simchen
Advances in Silicon Chemistry, 1, 189-189 null
E.J. Corey et al.
Tetrahedron Letters, 25, 4345-4345 (1984)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service