Skip to Content
Merck
All Photos(2)

Key Documents

917419

Sigma-Aldrich

2-(2-(Piperidin-4-yl)phenyl)ethan-1-amine dihydrochloride

Synonym(s):

Semi-flexible linker for PROTAC® development

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$908.00

$908.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
$908.00

About This Item

Empirical Formula (Hill Notation):
C13H22Cl2N2
Molecular Weight:
277.23
MDL number:
UNSPSC Code:
12352116
NACRES:
NA.22

$908.00


Check Cart for Availability

Request a Bulk Order

form

powder

Quality Level

reaction suitability

reagent type: linker

functional group

amine

storage temp.

2-8°C

SMILES string

NCCC1=C(C=CC=C1)C2CCNCC2.Cl.Cl

InChI

1S/C13H20N2.2ClH/c14-8-5-11-3-1-2-4-13(11)12-6-9-15-10-7-12;;/h1-4,12,15H,5-10,14H2;2*1H

InChI key

ONRWENIJVYXDCE-UHFFFAOYSA-N

Application

2-(2-(Piperidin-4-yl)phenyl)ethan-1-amine dihydrochloride is a 4-aryl piperidine useful as a semi-flexible linker in PROTAC development for targeted protein degradation. Incorporation of rigidity into the linker region of bifunctional protein degraders may impact the 3D orientation of the degrader and thus ternary complex formation as well as optimization of drug-like properties.

Legal Information

PROTAC is a registered trademark of Arvinas Operations, Inc., and is used under license

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service