Skip to Content
Merck
All Photos(4)

Key Documents

915246

Sigma-Aldrich

Grubbs catalyst® M203

Umicore

Synonym(s):

Tricyclohexylphosphine[1,3-bis(2,4,6-trimethylphenyl)imidazol-2-ylidene][3-phenyl-1H-inden-1-ylidene]ruthenium (II) dichloride, (IMes)Ru(PCy3)(Ind)Cl2, CatMETium RF1

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$318.00
1 G
$1,150.00

$318.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
100 MG
$318.00
1 G
$1,150.00

About This Item

Empirical Formula (Hill Notation):
C54H67Cl2N2PRu
CAS Number:
Molecular Weight:
947.07
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.22

$318.00


Estimated to ship on28 May 2025


Request a Bulk Order

Quality Level

form

powder

mp

230 °C (decomp)

SMILES string

CC1=CC(C)=CC(C)=C1n2c([Ru](Cl)(Cl)(P(C3CCCCC3)(C4CCCCC4)C5CCCCC5)=C6C(C=CC=C7)=C7C(C8=CC=CC=C8)=C6)n(C9=C(C)C=C(C)C=C9C)C=C2

InChI

1S/C21H24N2.C18H33P.C15H10.2ClH.Ru/c1-14-9-16(3)20(17(4)10-14)22-7-8-23(13-22)21-18(5)11-15(2)12-19(21)6;1-4-10-16(11-5-1)19(17-12-6-2-7-13-17)18-14-8-3-9-15-18;1-2-6-12(7-3-1)15-11-10-13-8-4-5-9-14(13)15;;;/h7-12H,1-6H3;16-18H,1-15H2;1-9,11H;2*1H;/q;;;;;+2/p-2

InChI key

HBNBIWQQLMQFEU-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Umicore Grubbs Catalyst M203 is a homogeneous catalyst useful for alkene metathesis, especially for cross-metathesis, ring-closing metathesis and self-metathesis.

Learn more about our metathesis catalysts

Storage and Stability

Do not store above 25°C (77°F). Store in a tightly closed original container under dry inert gas, protected from direct sunlight in a dry, cool and well-ventilated area.

Legal Information

Product of Umicore

Product License: This product, its manufacturing or use, is the subject of one or more issued or pending U.S. Patents (and foreign equivalents) owned or controlled by Umicore PMC. The purchase of this product from Umicore PMC through Sigma-Aldrich, its affiliates or their authorized distributors conveys to the buyer a limited, one-time, non-exclusive, non-transferable, non-assignable license. Buyer′s use of this product may infringe patents owned or controlled by third parties. It is the sole responsibility of buyer to ensure that its use of the product does not infringe the patent rights of third parties or exceed the scope of the license granted herein.

Additional information available at www.pmc.umicore.com
Grubbs Catalyst is a registered trademark of Umicore AG & Co. KG

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service