Skip to Content
Merck
All Photos(1)

Documents

91018

Supelco

(S)-NIFE

for chiral derivatization, ≥98.0%

Synonym(s):

N-(4-Nitrophenoxycarbonyl)-L-phenylalanine 2-methoxyethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H20N2O7
CAS Number:
Molecular Weight:
388.37
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

for chiral derivatization

Assay

≥98.0% (HPLC)
≥98.0%

form

powder

SMILES string

COCCOC(=O)[C@H](Cc1ccccc1)NC(=O)Oc2ccc(cc2)[N+]([O-])=O

InChI

1S/C19H20N2O7/c1-26-11-12-27-18(22)17(13-14-5-3-2-4-6-14)20-19(23)28-16-9-7-15(8-10-16)21(24)25/h2-10,17H,11-13H2,1H3,(H,20,23)/t17-/m0/s1

InChI key

ZSOUYIBYVUBOGT-KRWDZBQOSA-N

Other Notes

New chiral derivatizing agent for amino acid analysis by HPLC; derivatization of secondary amino acids

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sonika Sethi et al.
Biomedical chromatography : BMC, 34(1), e4730-e4730 (2019-10-28)
LC separation of biologically and pharmaceutically important enantiomers (from racemic or non-racemic mixtures) remains a subject of importance. The present review article deals with the liquid chromatographic enantioseparation of chiral selective serotonin reuptake inhibitors (SSRIs), namely citalopram, paroxetine, sertraline and
E. Olajos et al.
Chromatographia, 54, 77-77 (2001)
A. Peter et al.
Chromatographia, 52, 821-821 (2000)
Cecilia Barbas-Bernardos et al.
Journal of chromatography. A, 1611, 460598-460598 (2019-10-20)
In the nutrition field, there is a lack of understanding about the impact that dietary chiral composition may have on health, especially regarding cooked meals. Chiral amino acids (AAs) are naturally present in food and their proportion may vary quite
Antal Péter et al.
Journal of chromatography. A, 948(1-2), 283-294 (2003-07-02)
A new chiral derivatizing agent, (S)-N-(4-nitrophenoxycarbonyl)phenylalanine methoxyethyl ester, (S)-NIFE, was applied for the high-performance liquid chromatographic separation of enantiomers of 19 unnatural secondary amino acids: proline, pipecolic acid analogues, piperazine-2-carboxylic acid, morpholine-3-carboxylic acid, thiomorpholine-3-carboxylic acid and analogues containing the 1,2,3,4-tetrahydroisoquinoline

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service