Skip to Content
Merck
All Photos(1)

Documents

90919

Millipore

Enterococcus faecium ChromoSelect Agar Base

suitable for microbiology, NutriSelect® Plus

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171606
NACRES:
NA.85

sterility

non-sterile

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

composition

agar, 15.0 g/L
arabinose, 10.0 g/L
chromogenic substrate, 0.1 g/L
corn starch, 1.0 g/L
peptone, special, 23.0 g/L
phenol red, 0.1 g/L
sodium chloride, 5.0 g/L

manufacturer/tradename

NutriSelect® Plus

final pH

7.8±0.2 (25 °C)

application(s)

bioburden testing
clinical testing
environmental
food and beverages
water monitoring

microbiology

storage temp.

2-8°C

suitability

selective and differential for Enterococcus spp.

Application

Enterococcus faecium ChromoSelect Agar Baseis recommended for chromogenic identification of Enterococcus faecium from faeces, sewage and water supplies.

Preparation Note

Suspend 27.1 g in 500 ml distilled water. Heat to boiling to dissolve the medium completely. DO NOT AUTOCLAVE. Cool to 45-50°C and add the rehydrated contents of 1 vial of Enterococcus faecium Selective Supplement (Cat. No. 01318). Mix well and pour into sterile petri plates.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service