Skip to Content
Merck
All Photos(2)

Key Documents

907553

Sigma-Aldrich

(2R,3S,4S,5S,6R)-2-(((tert-butyldimethylsilyl)oxy)methyl)-6-(phenylthio)tetrahydro-2H-pyran-3,4,5-triol

≥95%

Synonym(s):

Phenyl 6-O-tert-butyldimemthylsilyl-1-thio-α-D-mannoside

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$607.00

$607.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
$607.00

About This Item

Empirical Formula (Hill Notation):
C18H30O5SSi
CAS Number:
Molecular Weight:
386.58
MDL number:
UNSPSC Code:
12352005
NACRES:
NA.22

$607.00


Check Cart for Availability

Request a Bulk Order

Assay

≥95%

form

chunks

storage temp.

2-8°C

SMILES string

[Si](OC[C@H]1O[C@@H]([C@H]([C@H]([C@@H]1O)O)O)Sc2ccccc2)(C(C)(C)C)(C)C

InChI key

TWBUWRKSGDZICZ-UHDSXZAQSA-N

Application

This is a protected mannopyranoside useful as a building block for synthesis of complex carbohydrates. The compound has α-phenylthio and 6-tert-butyldimethylsilyl protecting groups.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Carbohydrates and their biologically active glycoconjugate analogs are emerging as an important class of biomolecules.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service