Skip to Content
Merck
All Photos(1)

Documents

90467

Supelco

Cryptochlorogenic acid

analytical standard

Synonym(s):

4-O-Caffeoylquinic acid, 4-O-(3,4-Dihydroxycinnamoyl)-D-quinic acid, Cryptochlorogenic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H18O9
CAS Number:
Molecular Weight:
354.31
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤5.0% residual solvents
≤6.0% water

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

O[C@@H]1C[C@](O)(C[C@@H](O)[C@@H]1OC(=O)\C=C\c2ccc(O)c(O)c2)C(O)=O

InChI

1S/C16H18O9/c17-9-3-1-8(5-10(9)18)2-4-13(21)25-14-11(19)6-16(24,15(22)23)7-12(14)20/h1-5,11-12,14,17-20,24H,6-7H2,(H,22,23)/b4-2+/t11-,12-,14-,16+/m1/s1

InChI key

GYFFKZTYYAFCTR-JUHZACGLSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Cryptochlorogenic acid is a polyphenol, found in a variety of plants, which plays a crucial role in protection against various age-related diseases.

Application

Cryptochlorogenic acid may be used as a reference standard in the determination of cryptochlorogenic acid in the leaves of Hibiscus sabdariffa using high performance liquid chromatography coupled with mass spectrometry (HPLC-MS).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

LC/MS/MS characterization of phenolic constituents in dried plums.
Fang N, et al.
Journal of Agricultural and Food Chemistry, 50(12), 3579-3585 (2002)
Phytochemistry, antioxidant capacity, total phenolic content and anti-inflammatory activity of Hibiscus sabdariffa leaves.
Zhen J, et al.
Food Chemistry, 190, 673-680 (2016)
Liangmian Chen et al.
Chemical & pharmaceutical bulletin, 63(1), 25-32 (2015-03-07)
For the determination of seven caffeoylquinic acids [neochlorogenic acid (NcA), cryptochlorogenic acid (CcA), chlorogenic acid (CA), caffeic acid (CfA), isochlorogenic acid A (Ic A), isochlorogenic acid B (Ic B), isochlorogenic acid C (Ic C)] and two flavonoids [luteolin 7-O-glucoside (LtG) and luteolin (Lt)]
Liangmian Chen et al.
Journal of pharmaceutical and biomedical analysis, 95, 93-101 (2014-03-25)
A novel three-channel isocratic elution high performance liquid chromatography with electrochemical detection (3LC-ECD) system was developed for the simultaneous determination of various bioactive redox components in Shuang-Huang-Lian (SHL) preparations. This 3LC-ECD system consists of three isocratic elution flow ways, two

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service