Skip to Content
Merck
All Photos(1)

Key Documents

89928

Supelco

Azinphos-methyl-(dimethyl-d6)

PESTANAL®, analytical standard

Synonym(s):

Azinphos-methyl-d6, Methyltriazotin-d6

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$622.00

$622.00


Estimated to ship on24 April 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$622.00

About This Item

Empirical Formula (Hill Notation):
C10D6H6N3O3PS2
Molecular Weight:
323.36
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$622.00


Estimated to ship on24 April 2025


Request a Bulk Order

grade

analytical standard

Quality Level

product line

PESTANAL®

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

application(s)

agriculture

format

neat

storage temp.

2-8°C

SMILES string

O=C1N(CSP(OC([2H])([2H])[2H])(OC([2H])([2H])[2H])=S)N=NC2=CC=CC=C21

InChI

1S/C10H12N3O3PS2/c1-15-17(18,16-2)19-7-13-10(14)8-5-3-4-6-9(8)11-12-13/h3-6H,7H2,1-2H3/i1D3,2D3

InChI key

CJJOSEISRRTUQB-WFGJKAKNSA-N

General description

Azinphos-methyl-(dimethyl-d6) is an isotope-labeled analog of the organophosphorus anticholinesterase insecticide azinphos-methyl, wherein dimethoxy protons are substituted by deuterium.[1][2]

Application

Azinphos-methyl-(dimethyl-d6) may be used as a deuterated internal standard to determine the levels of pesticide analytes in foods,[3] fruits and vegetable matrices[4] by gas chromatography (GC) based techniques.
Isotope-labeled compounds are increasingly used in isotope dilution mass spectrometry (IDMS) for the quantitative analysis of pesticides. The major advantage of using this technique is that the isotope-labeled compounds have nearly the same physical properties as their non-labeled counterpart analogs, and thus show identical behavior during the workup and sample preparation process. This helps in overcoming the problems of matrix effects generally encountered in the usual LC-MS/GC-MS analysis potentially resulting in biased results. By spiking the sample before workup with its isotope labeled analog, the loss of analyte that leads to matrix effects can be determined and compensated.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 2 Oral - Acute Tox. 3 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effects of carbaryl and azinphos methyl on juvenile rainbow trout (Oncorhynchus mykiss) detoxifying enzymes
Ferrari A, et al.
Pesticide Biochemistry and Physiology, 88(2), 134-142 (2007)
Combined use of a modifier gas generator, analyte protectants and multiple internal standards for effective and robust compensation of matrix effects in gas chromatographic analysis of pesticides
Tsuchiyama T, et al.
Journal of Chromatography A, 1589(3), 122-133 (2019)
Compensation of matrix effects in gas chromatography-mass spectrometry analysis of pesticides using a combination of matrix matching and multiple isotopically labeled internal standards
Tsuchiyama T, et al.
Journal of Chromatography A, 1524(3), 233-245 (2017)
Ali Özcan et al.
Water research, 47(3), 1470-1479 (2013-01-02)
The removal of organophosphorous insecticide azinphos-methyl (AZPM) from water has been investigated by the electro-Fenton method which produces hydroxyl radicals electro-catalytically. The reaction between these radicals and AZPM has led to the oxidation of AZPM up to carbon dioxide and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service