Skip to Content
Merck
All Photos(1)

Key Documents

89189

Supelco

2-Methoxy-4-vinylphenol

analytical standard, contains 100 ppm BHT as inhibitor

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$86.00

$86.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
$86.00

About This Item

Linear Formula:
CH3OC6H3(CH=CH2)OH
CAS Number:
Molecular Weight:
150.17
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

$86.00


Check Cart for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥98.0% (GC)

shelf life

limited shelf life, expiry date on the label

contains

100 ppm BHT as inhibitor

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.582 (lit.)

bp

224 °C (lit.)

density

1.11 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

COc1cc(C=C)ccc1O

InChI

1S/C9H10O2/c1-3-7-4-5-8(10)9(6-7)11-2/h3-6,10H,1H2,2H3

InChI key

YOMSJEATGXXYPX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Varahenage R Perera et al.
Biochimica et biophysica acta, 1840(9), 2851-2861 (2014-05-14)
Gram-positive bacteria in the phylum Firmicutes synthesize the low molecular weight thiol bacillithiol rather than glutathione or mycothiol. The bacillithiol transferase YfiT from Bacillus subtilis was identified as a new member of the recently discovered DinB/YfiT-like Superfamily. Based on structural

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service