Skip to Content
Merck
All Photos(1)

Key Documents

87919

Sigma-Aldrich

Tetramethylrhodamine methyl ester perchlorate

for fluorescence, ≥95% (HPCE)

Synonym(s):

TMRM

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C25H25N2O3 · ClO4
CAS Number:
Molecular Weight:
500.93
Beilstein:
7162165
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:

grade

for fluorescence

Assay

≥95% (HPCE)

solubility

DMSO: soluble
alcohols: soluble

fluorescence

λex 540 nm; λem 595 nm in DMSO

suitability

suitable for fluorescence

storage temp.

2-8°C

SMILES string

[O-]Cl(=O)(=O)=O.COC(=O)c1ccccc1C2=C3C=C\C(C=C3Oc4cc(ccc24)N(C)C)=[N+](\C)C

InChI

1S/C25H25N2O3.ClHO4/c1-26(2)16-10-12-20-22(14-16)30-23-15-17(27(3)4)11-13-21(23)24(20)18-8-6-7-9-19(18)25(28)29-5;2-1(3,4)5/h6-15H,1-5H3;(H,2,3,4,5)/q+1;/p-1

InChI key

PFYWPQMAWCYNGW-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Potential-sensitive probe for measuring membrane potential changes in mitochondria

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Timothy M Tucey et al.
Cell metabolism, 27(5), 988-1006 (2018-05-03)
To fight infections, macrophages undergo a metabolic shift whereby increased glycolysis fuels antimicrobial inflammation and killing of pathogens. Here we demonstrate that the pathogen Candida albicans turns this metabolic reprogramming into an Achilles' heel for macrophages. During Candida-macrophage interactions intertwined
D L Farkas et al.
Biophysical journal, 56(6), 1053-1069 (1989-12-01)
The distribution of charged membrane-permeable molecular probes between intracellular organelles, the cytoplasm, and the outside medium is governed by the relative membrane electrical potentials of these regions through coupled equilibria described by the Nernst equation. A series of highly fluorescent

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service