Skip to Content
Merck
All Photos(1)

Key Documents

87253

Sigma-Aldrich

Tetraethylphosphonium hexafluorophosphate

≥98.0% (gravimetric)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2)4P(PF6)
CAS Number:
Molecular Weight:
292.18
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:

Assay

≥98.0% (gravimetric)

SMILES string

F[P-](F)(F)(F)(F)F.CC[P+](CC)(CC)CC

Other Notes

Novel organic electrolyte with high conductivity in high energy density electric double-layer capacitors (EDLC)

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T. Morimoto et al.
Proc. Electrochem. Soc., 93/23, 49-49 (1993)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service