Skip to Content
Merck
All Photos(1)

Key Documents

86899

Supelco

Tetrabutylammonium phosphate monobasic solution

suitable for ion pair chromatography, LiChropur, concentrate, ampule

Synonym(s):

Tetrabutylammonium dihydrogen phosphate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2CH2CH2)4N[OP(OH)2O]
CAS Number:
Molecular Weight:
339.45
Beilstein:
5196532
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NB.21

description

cationic

Quality Level

form

liquid

quality

LiChropur
ampule

packaging

ampule of 25 mL

technique(s)

ion pair chromatography: suitable

λ

in concentrate

UV absorption

λ: 210 nm Amax: 0.05
λ: 220 nm Amax: 0.04
λ: 300 nm Amax: 0.02
λ: 500 nm Amax: 0.02

suitability

corresponds to standard for RP gradient test
corresponds to standard for filter test

storage temp.

2-8°C

SMILES string

OP(O)([O-])=O.CCCC[N+](CCCC)(CCCC)CCCC

InChI

1S/C16H36N.H3O4P/c1-5-9-13-17(14-10-6-2,15-11-7-3)16-12-8-4;1-5(2,3)4/h5-16H2,1-4H3;(H3,1,2,3,4)/q+1;/p-1

InChI key

ARRNBPCNZJXHRJ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Concentrate for 6x1 L ready-to-use solution. Dilute contents of one ampule (~0.3 M) to 1 L with HPLC grade water to obtain a 0.005 M eluent solution.
Tetrabutylammonium dihydrogen phosphate is a quaternary ammonium salt mainly used as electrolyte.

Application

Tetrabutylammonium dihydrogen phosphate has been used as ion-pairing reagent in mobile phase for ion-pair HPLC analysis of 5′-mononucleotides from baby formulae.[1] It may also have been used in mobile phase for HPLC analysis of blood samples to determine acetaminophen.[2]

Other Notes

Ion-pairing reagent used in reversed phase chromatography [3][4][5]

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

In situ gelling and mucoadhesive liquid suppository containing acetaminophen: enhanced bioavailability.
Choi, Han-Gon, Yu-Kyoung Oh, and Chong-Kook Kim.
International Journal of Pharmaceutics, 165.1, 23-32 (1998)
W.E. Ladner et al.
The Journal of Organic Chemistry, 50, 1076-1076 (1985)
The analysis of 5'-mononucleotides in infant formulae by HPLC.
Perrin, Christian, et al.
Food Chemistry, 74.2, 245-253 (2001)
C.F. Harrington et al.
Journal of Liquid Chromatography and Related Technologies, 20, 1773-1773 (1997)
K Makino et al.
Journal of chromatography, 400, 271-277 (1987-07-29)
The separation of large oligodeoxyribonucleotides, synthesized chemically and subsequently deblocked, was carried out by reversed-phase ion-pair chromatography (RP-IPC) with a linear gradient of acetonitrile concentration. It was found that tetrabutylammonium phosphate is suitable as an ion-pairing reagent and produces a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service