Skip to Content
Merck
All Photos(1)

Key Documents

86851

Supelco

Tetrabutylammonium hydroxide solution

suitable for ion pair chromatography, concentrate, ampule

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2CH2CH2)4N(OH)
CAS Number:
Molecular Weight:
259.47
Beilstein:
3571228
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

quality

ampule

technique(s)

ion pair chromatography: suitable

λ

in concentrate

UV absorption

λ: 230 nm Amax: 0.10
λ: 240 nm Amax: 0.03
λ: 250 nm Amax: 0.02
λ: 500 nm Amax: 0.02

suitability

corresponds to standard for RP gradient test
corresponds to standard for filter test

storage temp.

2-8°C

SMILES string

[OH-].CCCC[N+](CCCC)(CCCC)CCCC

InChI

1S/C16H36N.H2O/c1-5-9-13-17(14-10-6-2,15-11-7-3)16-12-8-4;/h5-16H2,1-4H3;1H2/q+1;/p-1

InChI key

VDZOOKBUILJEDG-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Concentrate for 6x1 L ready-to-use solution. Dilute contents of one ampule (~0.3 M) to 1 L with HPLC grade water to obtain a 0.005 M eluent solution.

Physical form

can be solid (mp 25-30°C); warm up carefully for melting (max. 40°C)

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liwen Zhang et al.
ACS nano, 8(12), 12250-12258 (2014-11-18)
Photothermal therapy (PTT) is an emerging treatment modality that is under intensive preclinical investigations for the treatment of various medical conditions, including cancer. However, the lack of targeting function of PTT agents hampers its clinical application. An effective and nontoxic
Erbo Shi et al.
Organic letters, 14(13), 3384-3387 (2012-06-27)
A metal-free C-H oxidation for the construction of allylic esters has been developed. The use of a commercially available and inexpensive catalyst and oxidant, and readily available starting materials, coupled with the operational simplicity of the reaction, renders the methodology
Fabio Galeotti et al.
Journal of chromatography. A, 1284, 141-147 (2013-03-05)
In this study, by using tetrabutylammonium bisulfate as ion-pairing reagent, we were able to separate all the main heparin/heparan sulfate disaccharides generated by the action of heparinases along with the main Hep tetrasaccharide possessing a 3-O-sulfate group on the sulfoglucosamine
R Bryan Sears et al.
Journal of inorganic biochemistry, 121, 77-87 (2013-01-29)
The complex cis-[Ru(phpy)(phen)(CH3CN)2](+) (phpy=2-phenylpyridine, phen=1,10-phenanthroline) was investigated as a potential photodynamic therapy (PDT) agent. This complex presents desirable photochemical characteristics including a low energy absorption tail extending into the PDT window (600-850nm) and photoinduced exchange of the CH3CN ligands, generating
David J Posson et al.
Nature structural & molecular biology, 20(2), 159-166 (2012-12-25)
Understanding how ion channels open and close their pores is crucial for comprehending their physiological roles. We used intracellular quaternary ammonium blockers, electrophysiology and X-ray crystallography to locate the voltage-dependent gate in MthK potassium channels from Methanobacterium thermoautotrophicum. Blockers bind

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service