Skip to Content
Merck
All Photos(1)

Key Documents

861405

Sigma-Aldrich

Hoechst 33258

Synonym(s):

2′-(4-hydroxyphenyl)-5-(4-methyl-1-piperazinyl)-2,5′-bi-1H-benzimidazole trihydrochloride hydrate, bisBenzimide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H24N6O · 3HCl · xH2O
CAS Number:
Molecular Weight:
533.88 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:

Assay

≥98% (TLC)

form

powder

mp

>300 °C (lit.)

λmax

343 nm

SMILES string

Cl[H].Cl[H].Cl[H].[H]O[H].CN1CCN(CC1)c2ccc3[nH]c(nc3c2)-c4ccc5[nH]c(nc5c4)-c6ccc(O)cc6

Other Notes

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shawn N Whitehead et al.
PloS one, 6(6), e20808-e20808 (2011-06-21)
Gangliosides, a member of the glycosphingolipid family, are heterogeneously expressed in biological membranes and are particularly enriched within the central nervous system. Gangliosides consist of mono- or poly-sialylated oligosaccharide chains of variable lengths attached to a ceramide unit and are
Stud. Biophys., 125, 121-121 (1988)
Hélène Labit et al.
Nucleic acids research, 36(17), 5623-5634 (2008-09-04)
Replication origins in Xenopus egg extracts are located at apparently random sequences but are activated in clusters that fire at different times during S phase under the control of ATR/ATM kinases. We investigated whether chromosomal domains and single sequences replicate
Yi Liu et al.
Stem cells (Dayton, Ohio), 26(4), 1065-1073 (2008-02-02)
Periodontitis is a periodontal tissue infectious disease and the most common cause for tooth loss in adults. It has been linked to many systemic disorders, such as coronary artery disease, stroke, and diabetes. At present, there is no ideal therapeutic
G A Daxhelet et al.
Analytical biochemistry, 179(2), 401-403 (1989-06-01)
The increase in fluorescence, upon interaction with several fluorescent dyes was found to depend on the base composition of DNA. 4',6-Diamidino-2-phenylindole-2 HCl and Hoechst 33258 which bind to AT base pairs show a logarithmic relation. This relation is linear when

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service