Skip to Content
Merck
All Photos(1)

Key Documents

85427

Supelco

GC Stationary Phase

phase DC-710, bottle of 100 mL

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$410.00

$410.00


Estimated to ship on15 April 2025


Request a Bulk Order

Select a Size

Change View
100 ML
$410.00

About This Item

UNSPSC Code:
12000000
NACRES:
SB.52

$410.00


Estimated to ship on15 April 2025


Request a Bulk Order

form

liquid

Quality Level

packaging

bottle of 100 mL

technique(s)

gas chromatography (GC): suitable

refractive index

n20/D 1.537

matrix active group

DC-710 phase

viscosity

~500 mPa.s, neat(25 °C)

Looking for similar products? Visit Product Comparison Guide

General description

Synthesized specifically to be purer, of narrow molecular weight range, and without trace catalysts or impurities for use as a GC stationary phase.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service