Skip to Content
Merck
All Photos(1)

Key Documents

85030

Sigma-Aldrich

Serotonin creatinine sulfate salt monohydrate

purum, ≥98.0% (T)

Synonym(s):

5-Hydroxytryptamine creatinine sulfate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H12N2O · C4H9N3O5S · H2O
CAS Number:
Molecular Weight:
405.43
Beilstein:
3886526
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

purum

Assay

≥98.0% (T)

mp

~220 °C (dec.)

storage temp.

2-8°C

SMILES string

O.OS(O)(=O)=O.CN1CC(=O)NC1=N.NCCc2c[nH]c3ccc(O)cc23

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Linda Booij et al.
PloS one, 10(3), e0119061-e0119061 (2015-03-18)
Serotonin plays an important role in the etiology of depression. Serotonin is also crucial for brain development. For instance, animal studies have demonstrated that early disruptions in the serotonin system affect brain development and emotion regulation in later life. A
Di Wang et al.
Journal of neurophysiology, 112(1), 30-38 (2014-04-18)
The lamprey central nervous system has been used extensively as a model system for investigating the networks underlying vertebrate motor behavior. The locomotor networks can be activated by application of glutamate agonists, such as N-methyl-D-aspartic acid (NMDA), to the isolated
Toko Maehara et al.
British journal of pharmacology, 172(4), 1136-1147 (2014-11-08)
Post-operative ileus (POI) is induced by intestinal inflammation. Here, we aimed to clarify the effects of 5-HT3 receptor antagonists against POI. We administered three 5-HT3 receptor antagonists, ondansetron, tropisetron and palonosetron, to a mouse model of POI induced by surgical
Mathieu Desclaux et al.
Journal of neuroscience research, 93(1), 43-55 (2014-08-19)
In spinal cord injury (SCI), absence of functional recovery and lack of spontaneous axonal regeneration are attributed, among other factors, to the formation of a glial scar that forms both physical and chemical barriers. The glial scar is composed mainly
Kirsty Bannister et al.
Neuropharmacology, 85, 375-383 (2014-06-18)
Dynorphin A is an endogenous opioid peptide derived from the precursor prodynorphin. The proteolytic fragment dynorphin A (1-17) exhibits inhibitory effects via opioid receptors. Paradoxically, the activity of the dynorphin system increases with chronic pain and neuropathy is associated with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service