Skip to Content
Merck
All Photos(1)

Key Documents

83672

Supelco

Rhenium standard for AAS

analytical standard, ready-to-use, in nitric acid

Synonym(s):

Rhenium nitrate solution

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$104.00

$104.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
100 ML
$104.00

About This Item

MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

$104.00


Estimated to ship on28 May 2025


Request a Bulk Order

grade

analytical standard

shelf life

limited shelf life, expiry date on the label

concentration

in nitric acid
1.000 g/L Re

technique(s)

AAS: suitable

format

single component solution

SMILES string

[ReH]

InChI

1S/Re

InChI key

WUAPFZMCVAUBPE-UHFFFAOYSA-N

Preparation Note

prepared with Re in HNO3 (~1 M)

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service