Skip to Content
Merck
All Photos(1)

Key Documents

82330

Sigma-Aldrich

Propylene sulfide

purum, ≥96.0% (GC)

Synonym(s):

Methylthiirane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H6S
CAS Number:
Molecular Weight:
74.14
Beilstein:
383559
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

purum

Assay

≥96.0% (GC)

refractive index

n20/D 1.475 (lit.)
n20/D 1.475

bp

72-75 °C (lit.)

density

0.946 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CC1CS1

InChI

1S/C3H6S/c1-3-2-4-3/h3H,2H2,1H3

InChI key

MBNVSWHUJDDZRH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Inhalation - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

50.0 °F - closed cup

Flash Point(C)

10 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cong-Duan Vo et al.
Macromolecular rapid communications, 34(2), 156-162 (2013-01-16)
We report for the first time the combination of ATRP and ring-opening episulfide polymerization as a means to synthesize polysulfide-based low-dispersity amphiphilic block copolymers. The most significant finding is the possibility to perform ATRP under mild conditions using poly(propylene sulfide)
Masamichi Yamanaka et al.
Chemical communications (Cambridge, England), (15)(15), 1690-1691 (2004-07-28)
Three chiral guests in a cylindrical capsule led to six diastereomeric complexes.
J P Bearinger et al.
Nature materials, 2(4), 259-264 (2003-04-12)
Alkanethiolates have been widely used as chemisorbates to modify gold surfaces, in spite of their relatively poor oxidative stability. We introduce gold-chemisorbing block copolymers bearing an anchoring block of poly(propylene sulphide) (PPS), selected in the expectation of greater stability. These
T Matsuda et al.
Cancer letters, 86(2), 229-234 (1994-11-11)
Two organosulfur compounds, methyl propyl disulfide (MPD) and propylene sulfide (PS) from garlic and onions, were studied for their modifying effects on hepatocarcinogenesis in the F344 rats. Modifying potential was scored by comparing the number and area per cm2 of
Biomaterials: Stable antifouling surfaces.
Ralph G Nuzzo
Nature materials, 2(4), 207-208 (2003-04-12)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service