Skip to Content
Merck
All Photos(1)

Key Documents

81572

Sigma-Aldrich

5α-Pregnan-3α-ol-20-one

≥98.0% (TLC)

Synonym(s):

3α-Hydroxy-5α-pregnan-20-one, 3α-OH-DHP, Allopregnan-3α-ol-20-one, Allopregnanolone, Brexanolone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H34O2
CAS Number:
Molecular Weight:
318.49
Beilstein:
3211363
MDL number:
UNSPSC Code:
12352200

Assay

≥98.0% (TLC)

solubility

chloroform: 20 mg/mL, clear, colorless

SMILES string

[H][C@@]12CC[C@@]3([H])[C@]4([H])CC[C@H](C(C)=O)[C@@]4(C)CC[C@]3([H])[C@@]1(C)CC[C@@H](O)C2

InChI

1S/C21H34O2/c1-13(22)17-6-7-18-16-5-4-14-12-15(23)8-10-20(14,2)19(16)9-11-21(17,18)3/h14-19,23H,4-12H2,1-3H3/t14-,15+,16-,17+,18-,19-,20-,21+/m0/s1

InChI key

AURFZBICLPNKBZ-SYBPFIFISA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cristina Suñol et al.
Journal of medicinal chemistry, 49(11), 3225-3234 (2006-05-26)
A GABA(A) receptor study of several B-nor analogues of allopregnanolone and pregnanolone has been carried out. B-norallopregnanolone (i.e., 3alpha-hydroxy-7-nor-5alpha-pregnan-20-one) was found comparable to allopregnanolone when measured with labeled TBPS. Analogous results were obtained from their effect on neurons in culture:
Samira Afrazi et al.
The Journal of steroid biochemistry and molecular biology, 139, 98-103 (2013-11-02)
Hyperglycemia plays a critical role in the development of diabetic neuropathy. Hyperglycemia induces oxidative stress in neurons, resulting in neuronal cell apoptosis and dysfunction. Anti-apoptotic properties of neurosteroids have been demonstrated in numerous cellular models of neurodegenerative studies. Here, the
Jonathan J Hirst et al.
The Journal of steroid biochemistry and molecular biology, 139, 144-153 (2013-05-15)
Neuroactive steroid concentrations are remarkably high in the foetal brain during late gestation. These concentrations are maintained by placental progesterone synthesis and the interaction of enzymes in the placenta and foetal brain. 5α-Pregnane-3α-ol-20-one (allopregnanolone) is a key neuroactive steroid during
Samuel O Adeosun et al.
PloS one, 7(11), e50040-e50040 (2012-12-05)
Restorative/protective therapies to restore dopamine neurons in the substantia nigra pars compacta (SNpc) are greatly needed to effectively change the debilitating course of Parkinson's disease. In this study, we tested the therapeutic potential of a neurogenic neurosteroid, allopregnanolone, in the
Jane Evans et al.
Neuropharmacology, 63(8), 1315-1326 (2012-09-04)
Chronic stress has been implicated as a causal factor in depression and anxiety, and is associated with neuroendocrine dysfunction and impaired hippocampal neurogenesis. The neurosteroid allopregnanolone (3α,5α-THP; ALLO) has been shown to be reduced in depressed patients. ALLO is "stress

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service