Skip to Content
Merck
All Photos(1)

Key Documents

80630

Supelco

Piperazine hexahydrate

≥98.0% (T)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H10N2 · 6H2O
CAS Number:
Molecular Weight:
194.23
Beilstein:
4455527
EC Number:
MDL number:
UNSPSC Code:
12000000

vapor pressure

0.8 mmHg ( 20 °C)

Assay

≥98.0% (T)

bp

145-156 °C (lit.)

mp

42-44 °C (lit.)
42-44 °C

SMILES string

[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].C1CNCCN1

InChI

1S/C4H10N2.6H2O/c1-2-6-4-3-5-1;;;;;;/h5-6H,1-4H2;6*1H2

InChI key

AVRVZRUEXIEGMP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mitsunori Kono et al.
Bioorganic & medicinal chemistry, 21(1), 28-41 (2012-12-12)
A series of piperazine ureas was designed, synthesized, and evaluated for their potential as novel orally available fatty acid amide hydrolase (FAAH) inhibitors that are therapeutically effective against pain. We carried out an optimization study of the lead compound 3
William Conway et al.
The journal of physical chemistry. A, 117(5), 806-813 (2013-01-05)
Piperazine (PZ) is widely recognized as a promising solvent for postcombustion capture (PCC) of carbon dioxide (CO(2)). In view of the highly conflicting data describing the kinetic reactions of CO(2)(aq) in piperazine solutions, the present study focuses on the identification
Yuji Nakamura et al.
Bioorganic & medicinal chemistry letters, 22(14), 4561-4566 (2012-06-26)
Introduction of the 2,2-dimethyl-4-phenylpiperazin-5-one scaffold into the P(3)-P(1) portion of the (2S,4S,5S)-5-amino-6-dialkylamino-4-hydroxy-2-isopropylhexanamide backbone dramatically increased the renin inhibitory activity without using the interaction to the S(3)(sp) pocket. Compound 31 exhibited >10,000-fold selectivity over other human proteases, and 18.5% oral bioavailability
John A Christopher et al.
Journal of medicinal chemistry, 56(9), 3446-3455 (2013-03-23)
Biophysical fragment screening of a thermostabilized β1-adrenergic receptor (β1AR) using surface plasmon resonance (SPR) enabled the identification of moderate affinity, high ligand efficiency (LE) arylpiperazine hits 7 and 8. Subsequent hit to lead follow-up confirmed the activity of the chemotype
Enza Lacivita et al.
Journal of medicinal chemistry, 55(14), 6375-6380 (2012-06-29)
Here we report the design, synthesis, and 5-HT(7) receptor affinity of a set of 1-(3-biphenyl)- and 1-(2-biphenyl)piperazines. The effect on 5-HT(7) affinity of various substituents on the second (distal) phenyl ring was analyzed. Several compounds showed 5-HT(7) affinities in the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service