Skip to Content
Merck
All Photos(1)

Documents

80547

Millipore

Rhamnose Broth

suitable for microbiology, NutriSelect® Basic

Synonym(s):

LRB

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171606
NACRES:
NA.74

Agency

according to ISO 11290-1:2017
according to ISO 11290-2:2017

Quality Level

sterility

non-sterile

form

powder

shelf life

limited shelf life, expiry date on the label

composition

bromocresol purple, 0.02 g/L
meat extract, 1 g/L
peptone from meat, 10 g/L
sodium chloride, 5 g/L

manufacturer/tradename

NutriSelect® Basic

technique(s)

microbiological culture: suitable

final pH

6.8±0.2 (25 °C)

application(s)

agriculture
environmental
environmental
food and beverages
food and beverages

microbiology

suitability

nonselective and differential for Listeria spp.

Application

For the biochemical differentiation of Listeria subspecies. Is also base media for the Methyl-alpha-D-mannopyranoside Broth.

Preparation Note

Supplements :
100 ml/l Rhamnose Broth Supplement (Cat. No. 80301) or
100 ml/l Methyl-α-D-mannopyranoside Supplement (Cat. No. 02046)

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service