Skip to Content
Merck
All Photos(1)

Key Documents

80452

Sigma-Aldrich

Picric acid

purum, moistened with ≥40% water, ≥98.0% (calc. on dry substance, T)

Synonym(s):

2,4,6-Trinitrophenol, PA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(O2N)3C6H2OH
CAS Number:
Molecular Weight:
229.10
Colour Index Number:
10305
Beilstein:
423400
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

purum

Assay

≥98.0% (calc. on dry substance, T)

form

solid

quality

moistened with ≥40% water

expl. lim.

0.01 %

impurities

≥40% water

mp

122-123 °C (dried material) (lit.)

SMILES string

Oc1c(cc(cc1[N+]([O-])=O)[N+]([O-])=O)[N+]([O-])=O

InChI

1S/C6H3N3O7/c10-6-4(8(13)14)1-3(7(11)12)2-5(6)9(15)16/h1-2,10H

InChI key

OXNIZHLAWKMVMX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Desen. Expl. 1

Storage Class Code

4.1A - Other explosive hazardous materials

WGK

WGK 2

Flash Point(F)

302.0 °F - closed cup

Flash Point(C)

150 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Giuliana Magri et al.
Nature immunology, 15(4), 354-364 (2014-02-25)
Innate lymphoid cells (ILCs) regulate stromal cells, epithelial cells and cells of the immune system, but their effect on B cells remains unclear. Here we identified RORγt(+) ILCs near the marginal zone (MZ), a splenic compartment that contains innate-like B
Zhonghua Xiang et al.
Macromolecular rapid communications, 33(14), 1184-1190 (2012-04-18)
Three porous luminescent covalent--organic polymers (COPs) have been synthesized through self-polycondensation of the monomers of tris(4-bromophenyl)amine, 1,3,5-tris(4-bromophenyl)benzene, and 2,4,6-tris-(4-bromo-phenyl)-[1,3,5]triazine by using Ni-catalyzed Yamamoto reaction. All the COP materials possess not only high Brunauer-Emmett-Teller (BET) specific surface area of about 2000
Debby Kruijsen et al.
Journal of virology, 87(13), 7550-7557 (2013-05-03)
Infants are protected from a severe respiratory syncytial virus (RSV) infection in the first months of life by maternal antibodies or by prophylactically administered neutralizing antibodies. Efforts are under way to produce RSV-specific antibodies with increased neutralizing capacity compared to
Yingxin Ma et al.
Analytical chemistry, 84(19), 8415-8421 (2012-09-06)
Rapid, sensitive, and selective detection of explosives such as 2,4,6-trinitrotoluene (TNT) and 2,4,6-trinitrophenol (TNP), especially using a facile paper sensor, is in high demand for homeland security and public safety. Although many strategies have been successfully developed for the detection
Dmitry Momotenko et al.
Analytical chemistry, 84(15), 6630-6637 (2012-07-14)
This manuscript presents a push-pull electrochemical scanner able to image reactivity of initially dry surfaces by scanning electrochemical microscopy (SECM) and to probe molecules present or generated at the surface by mass spectrometry (MS). The proof-of-concept is demonstrated by coupling

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service