80380624-0010-P
Europium Functionalized Microspheres
F1-EU 030-0.1G-P, polystyrene, carboxylic acid functionalized, europium labeled, vial of 0.1 g, fluorescent red
Select a Size
$984.00
Estimated to ship on30 April 2025
Select a Size
About This Item
$984.00
Estimated to ship on30 April 2025
Recommended Products
label
europium labeled
material
polyethylene bottle
polystyrene
packaging
vial of 0.1 g
manufacturer/tradename
Estapor®
concentration
1% solids
color
fluorescent red
diameter
0.270-0.330 μm
capacity
200-500 μeq/g surface charge density
functional group
carboxylic acid
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
General description
Significantly improve lateral flow assay sensitivity.
- Drastically reduce background fluorescence.
- Make assays easier to read and quantify.
- Exhibit a much longer Stokes shift than traditional fluorescent labels.
- Exhibit an enhanced fluorescent quantum yield facilitating a low detection limit.
- Are functionalized with a carboxylated surface for protein conjugation.
- Are available in three different size options.
Application
Legal Information
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service