Skip to Content
Merck
All Photos(2)

Key Documents

80286

Supelco

2,4,6-Tris(trifluoromethyl)-1,3,5-triazine

analytical standard, suitable for mass spectrometry (MS)

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$88.80

$88.80


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$88.80

About This Item

Empirical Formula (Hill Notation):
C6F9N3
CAS Number:
Molecular Weight:
285.07
Beilstein:
302339
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$88.80


Check Cart for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥98.5% (GC)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable
mass spectrometry (MS): suitable

refractive index

n20/D 1.313 (lit.)

bp

98.3-98.5 °C/748 mmHg (lit.)

application(s)

environmental

format

neat

storage temp.

2-8°C

SMILES string

FC(F)(F)c1nc(nc(n1)C(F)(F)F)C(F)(F)F

InChI

1S/C6F9N3/c7-4(8,9)1-16-2(5(10,11)12)18-3(17-1)6(13,14)15

InChI key

LSGBKABSSSIRJF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service