Skip to Content
Merck
All Photos(1)

Key Documents

792780

Sigma-Aldrich

Ethyl alcohol, Pure

≥99.5%, ACS reagent, 200 proof, meets USP testing specifications, Excise Tax-free, Permit for use required

Synonym(s):

Tax-free ethanol, Ethanol, absolute alcohol, non-denatured ethanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2OH
CAS Number:
Molecular Weight:
46.07
EC Number:
MDL number:
UNSPSC Code:
15101802
PubChem Substance ID:
grade:
ACS reagent
application(s):
cleaning products
clinical research
cosmetics
flavors and fragrances
food and beverages
life science and biopharma
personal care
bp:
78 °C

Product Name

Ethyl alcohol, Pure, 200 proof, ACS reagent, meets USP testing specifications, Excise Tax-free, Permit for use required

grade

ACS reagent

Quality Level

Agency

meets USP testing specifications

vapor density

1.59 (vs air)

Assay

≥99.5%

form

liquid

expl. lim.

3.1-27.7 %

availability

not available in EU

concentration

200 proof

dilution

(for analytical testing)

impurities

≤0.2% (water)

color

colorless

pH

7 (20 °C, 10 g/L)

bp

78 °C

mp

-143.99 °C (-227.18 °F)

application(s)

cleaning products
clinical research
cosmetics
flavors and fragrances
food and beverages
life science and biopharma
personal care

Tax and license information

Excise tax-free with license
Permit for use required

SMILES string

OCC

InChI

1S/C2H6O/c1-2-3/h3H,2H2,1H3

InChI key

LFQSCWFLJHTTHZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Useful for multiple research applications in universities and hospitals.

Packaging

24x1PT is in glass bottle.
24x1PT-PB is in poly bottle.
72x1PT is 3x(24x1PT) in glass bottle.
72x1PT-PB is 3x(24x1PT) in poly bottle.
120x1PT is 5x(24x1PT) in glass bottle.
120x1PT-PB is 5x(24x1PT) in poly bottle.
4x1GA is in glass bottle.
4x1GA-PB is in poly bottle.
12x1GA is 3x(4x1GA) in glass bottle.
12x1GA-PB is 3x(4x1GA) in poly bottle.
20x1GA is 5x(4x1GA) in glass bottle.
20x1GA-PB is 5x(4x1GA) in poly bottle.
5GA is in metal drum with flex spout.
5GA-PB is in a poly drum with flex spout.
5GA-CU is in a cubetainer.
3x5GA is in metal drum with flex spout.
3x5GA-PB is in a poly drum with flex spout.
3x5GA-CU is in a cubetainer.
55GA is in metal drum.
55GA-PB is in poly drum.

Other Notes

Ordering Information:
To purchase tax-free ethyl alcohol, customers must possess an Industrial Alcohol User Permit from the Alcohol and Tobacco Tax and Trade Bureau (TTB) specifically designated for the withdrawal and use of alcohol free of tax.
  • Permit must have customer signature and date of submission to Sigma-Aldrich in addition to the original signature and date.
  • Permit must have word, "copy" handwritten or stamped.
  • Permit number must contain correct state or US designation matching ship-to address.
  • Permit address must match ship-to-address in Sigma-Aldrich system.
  • Permit only needs to be provided on first order to [email protected].
  • All orders require that a PO with the statement, "valid permit #####" be submitted to [email protected].

TTB Link

related product

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

55.4 °F - closed cup

Flash Point(C)

13 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ross B Mounsey et al.
Experimental neurology, 273, 36-44 (2015-08-06)
Parkinson's disease (PD) is a common chronic neurodegenerative disorder, usually of idiopathic origin. Symptoms including tremor, bradykinesia, rigidity and postural instability are caused by the progressive loss of dopaminergic neurons in the nigrostriatal region of the brain. Symptomatic therapies are
N Di Girolamo et al.
Stem cells (Dayton, Ohio), 33(1), 157-169 (2014-06-27)
Stem cell (SC) division, deployment, and differentiation are processes that contribute to corneal epithelial renewal. Until now studying the destiny of these cells in a living mammal has not been possible. However, the advent of inducible multicolor genetic tagging and
Cem Kuscu et al.
Nature biotechnology, 32(7), 677-683 (2014-05-20)
RNA-guided genome editing with the CRISPR-Cas9 system has great potential for basic and clinical research, but the determinants of targeting specificity and the extent of off-target cleavage remain insufficiently understood. Using chromatin immunoprecipitation and high-throughput sequencing (ChIP-seq), we mapped genome-wide
Rubaiyea Farrukee et al.
The Journal of antimicrobial chemotherapy, 70(7), 2004-2012 (2015-03-20)
The burden of disease due to influenza B is often underestimated. Clinical studies have shown that oseltamivir, a widely used neuraminidase inhibitor (NAI) antiviral drug, may have reduced effectiveness against influenza B viruses. Therefore, it is important to study the
Stefan Wieland et al.
Hepatology (Baltimore, Md.), 59(6), 2121-2130 (2013-10-15)
Approximately 50% of patients with chronic hepatitis C (CHC) have ongoing expression of interferon stimulated genes (ISGs) in the liver. It is unclear why this endogenous antiviral response is inefficient in eradicating the infection. Several viral escape strategies have been

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service