Skip to Content
Merck
All Photos(1)

Key Documents

79213

Supelco

1,1′-Oxybis[3-(2-methoxyphenoxy)-2-propanol]

pharmaceutical impurity standard

Synonym(s):

Guaifenesin impurity C (PhEur)

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
$1,440.00

$1,440.00


Estimated to ship on01 June 2025


Request a Bulk Order

Select a Size

Change View
50 MG
$1,440.00

About This Item

Empirical Formula (Hill Notation):
C20H26O7
Molecular Weight:
378.42
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$1,440.00


Estimated to ship on01 June 2025


Request a Bulk Order

Quality Level

API family

guaifenesin

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

impurities

≤5.0% water (calc. from elemental analysis)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

OC(COCC(COC1=C(OC)C=CC=C1)O)COC2=CC=CC=C2OC

InChI

1S/C20H26O7/c1-23-17-7-3-5-9-19(17)26-13-15(21)11-25-12-16(22)14-27-20-10-6-4-8-18(20)24-2/h3-10,15-16,21-22H,11-14H2,1-2H3

InChI key

AFKSAYSHFVIMCI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service