Skip to Content
Merck
All Photos(1)

Key Documents

79184BC

Millipore

GDS variable Amp Tube Lid, 72 well

BioControl Systems, for use with GDS Rotor-Gene®

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
$512.00

$512.00


Estimated to ship on07 May 2025


Request more information

Select a Size

Change View
1 EACH
$512.00

About This Item

UNSPSC Code:
41104017
NACRES:
NA.85

$512.00


Estimated to ship on07 May 2025


Request more information

manufacturer/tradename

BioControl Systems

technique(s)

PCR: suitable

application(s)

food and beverages
pathogen testing

compatibility

for use with GDS Rotor-Gene®

storage temp.

room temp

Legal Information

Rotor-Gene is a registered trademark of Qiagen GmbH

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service