78682
L-Serine
tested according to Ph. Eur.
Synonym(s):
Serinum, (S)-2-Amino-3-hydroxypropionic acid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Agency
USP/NF
tested according to Ph. Eur.
form
powder
mp
222 °C (dec.) (lit.)
solubility
H2O: very soluble 359.7 g/L at 20 °C, clear (OECD Test Guideline 105)
SMILES string
N[C@@H](CO)C(O)=O
InChI
1S/C3H7NO3/c4-2(1-5)3(6)7/h2,5H,1,4H2,(H,6,7)/t2-/m0/s1
InChI key
MTCFGRXMJLQNBG-REOHCLBHSA-N
Looking for similar products? Visit Product Comparison Guide
Biochem/physiol Actions
Precursor of glycine by serine hydroxymethyltransferase.
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Science (New York, N.Y.), 343(6169), 408-411 (2014-01-25)
Plant cells are immobile; thus, plant growth and development depend on cell expansion rather than cell migration. The molecular mechanism by which the plasma membrane initiates changes in the cell expansion rate remains elusive. We found that a secreted peptide
Progress in neuro-psychopharmacology & biological psychiatry, 29(5), 767-769 (2005-06-09)
Several lines of evidence suggest that D-serine, an endogenous agonist of the glycine site on the NMDA receptors, might play a role in the pathophysiology of schizophrenia. The purpose of this study was to determine whether levels of D- and
Nucleic acids research, 41(12), 6058-6071 (2013-05-02)
To fit within the confines of the cell, bacterial chromosomes are highly condensed into a structure called the nucleoid. Despite the high degree of compaction in the nucleoid, the genome remains accessible to essential biological processes, such as replication and
Science (New York, N.Y.), 344(6185), 760-764 (2014-05-17)
A switchlike response in nuclear factor-κB (NF-κB) activity implies the existence of a threshold in the NF-κB signaling module. We show that the CARD-containing MAGUK protein 1 (CARMA1, also called CARD11)-TAK1 (MAP3K7)-inhibitor of NF-κB (IκB) kinase-β (IKKβ) module is a
The Biochemical journal, 371(Pt 3), 653-661 (2003-01-22)
The amino acid L-serine, one of the so-called non-essential amino acids, plays a central role in cellular proliferation. L-Serine is the predominant source of one-carbon groups for the de novo synthesis of purine nucleotides and deoxythymidine monophosphate. It has long
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service