Skip to Content
Merck
All Photos(2)

Documents

78411

Sigma-Aldrich

o-Phenylenediamine

for fluorescence, free base

Synonym(s):

1,2-Diaminobenzene, 1,2-Phenylenediamine, OPD

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8N2
CAS Number:
Molecular Weight:
108.14
Beilstein:
606074
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

grade

for fluorescence

vapor density

3.7 (vs air)

vapor pressure

0.01 mmHg ( 25 °C)

form

tablet

concentration

20 mg/tablet (~45 mg)

bp

256-258 °C

mp

100-102 °C

solubility

H2O: 1 tablet/10 mL, clear, colorless

storage temp.

2-8°C

SMILES string

Nc1ccccc1N

InChI

1S/C6H8N2/c7-5-3-1-2-4-6(5)8/h1-4H,7-8H2

InChI key

GEYOCULIXLDCMW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Oxidation to 2,3-diaminophenazine; Determination of urinary oxalate by HPLC; Fluorescent detection of pyruvate

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Irrit. 2 - Muta. 2 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

276.8 °F - closed cup

Flash Point(C)

136 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of urinary oxalate by high-performance liquid chromatography.
H Hughes et al.
Analytical biochemistry, 119(1), 1-3 (1982-01-01)
L.D. Buckberry et al
Journal of Liquid Chromatography, 16, 3073-3073 (1993)
P J Tarcha et al.
Analytical biochemistry, 165(1), 230-233 (1987-08-15)
NMR and mass spectroscopic evidence has been obtained which indicates that the product of the oxidation of o-phenylenediamine by hydrogen peroxide, uncatalyzed or catalyzed by horseradish peroxidase, is 2,3-diaminophenazine. These results settle disparate literature descriptions. The process is most likely
Hai-Long Wang et al.
Acta tropica, 137, 58-66 (2014-05-13)
Nasal vaccination is an effective therapeutic regimen for preventing certain infectious diseases. The mucosal immune response is important for resistance to Toxoplasma gondii infection. In this study, we evaluated the immune responses elicited in BALB/c mice by nasal immunisation with
Robert D Inman et al.
Arthritis research & therapy, 11(1), R19-R19 (2009-02-11)
We have previously observed that Brown Norway (BN) rats display a relative resistance to experimental Chlamydia-induced arthritis. In the present study, we examine an environmental toxin, mercuric chloride (HgCl2), as a modulator of this innate resistance to arthritis. To assess

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service