Skip to Content
Merck
All Photos(1)

Key Documents

78268

Supelco

N-Methyl-bis-heptafluorobutyramide

for GC derivatization, LiChropur, ≥96.0% (GC)

Synonym(s):

N-Methyl-bis-perfluorobutyramide, MBHFBA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H3F14NO2
CAS Number:
Molecular Weight:
423.10
Beilstein:
6986874
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NA.22

grade

derivatization grade ((GC derivatization))
for GC derivatization

Quality Level

Assay

≥96.0% (GC)

form

liquid

quality

LiChropur

reaction suitability

reagent type: derivatization reagent
reaction type: Acylations

technique(s)

gas chromatography (GC): suitable

refractive index

n20/D 1.323

density

1.686 g/mL at 20 °C (lit.)

SMILES string

CN(C(=O)C(F)(F)C(F)(F)C(F)(F)F)C(=O)C(F)(F)C(F)(F)C(F)(F)F

InChI

1S/C9H3F14NO2/c1-24(2(25)4(10,11)6(14,15)8(18,19)20)3(26)5(12,13)7(16,17)9(21,22)23/h1H3

InChI key

BCQYNSPESDDJBZ-UHFFFAOYSA-N

General description

N-Methyl-bis-heptafluorobutyramide is an acylation reagent for GC/MS derivatization with benefits:
  • Improves stability of compounds by protecting unstable groups.
  • It may confer volatility on substances such as carbohydrates or amino acids, which have so many polar groups that they are non-volatile and normally decompose on heating.
  • It assists in separations not possible with underivatized compounds.
  • Compounds are detectable at very low levels with an ECD.

Other Notes

Derivatizing agent for amines, alcohols and thiols, forming neutral butyramide

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

V Cirimele et al.
Journal of chromatography. B, Biomedical sciences and applications, 740(2), 265-271 (2000-05-23)
A sensitive, specific and reproducible method for the quantitative determination of stanozolol in human hair has been developed. The sample preparation involved a decontamination step of the hair with methylene chloride and the sonication in methanol of 100 mg of
Alvaro Aragon et al.
Analytical and bioanalytical chemistry, 409(11), 2931-2939 (2017-02-17)
A sensitive analytical method for the determination of monoamine neurotransmitters (MNTs) in zebrafish larvae was developed using gas chromatography coupled to mass spectrometry. Six MNTs were selected as target compounds for neurotoxicity testing. MNTs underwent a two-step derivatization with hexamethyldisilazane
Franziska Gaunitz et al.
International journal of legal medicine, 132(6), 1645-1657 (2018-09-17)
This case report centres on the post-mortem distribution of the synthetic cannabinoid MDMB-CHMICA and its metabolites in the case of a 27-year-old man found dead after falling from the 24th floor of a high-rise building. Toxicological analysis of post-mortem samples
Hyun Ju Shin et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 1051, 97-107 (2017-03-25)
The tyrosine, tryptophan, and glutamate metabolic pathways play key roles on pathological state of neuronal functions and the change of their levels in biological systems reflects the progress degree of neuronal diseases. Comprehensive profiling of these metabolites is important to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service