Skip to Content
Merck
All Photos(1)

Key Documents

78047

Supelco

3-Hydroxy-DL-kynurenine

analytical standard

Synonym(s):

α,2-Diamino-3-hydroxy-γ-oxobenzenebutanoic acid

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$183.00

$183.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
10 MG
$183.00

About This Item

Empirical Formula (Hill Notation):
C10H12N2O4
CAS Number:
Molecular Weight:
224.21
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

$183.00


Estimated to ship on28 May 2025


Request a Bulk Order

grade

analytical standard

Assay

≥90.0% (HPLC)

form

powder

shelf life

limited shelf life, expiry date on the label

application(s)

clinical testing

storage temp.

2-8°C

SMILES string

NC(CC(=O)c1cccc(O)c1N)C(O)=O

InChI

1S/C10H12N2O4/c11-6(10(15)16)4-8(14)5-2-1-3-7(13)9(5)12/h1-3,6,13H,4,11-12H2,(H,15,16)

InChI key

VCKPUUFAIGNJHC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service