Skip to Content
Merck
All Photos(1)

Key Documents

76750

Supelco

Chlorodimethyl(pentafluorophenyl)silane

for GC derivatization, LiChropur, ≥95.0%

Synonym(s):

(Pentafluorophenyl)dimethylchlorosilane, Flophemesyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6F5Si(CH3)2Cl
CAS Number:
Molecular Weight:
260.66
Beilstein:
2858407
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NA.22

grade

for GC derivatization

Quality Level

Assay

≥95.0% (GC)
≥95.0%

form

liquid

quality

LiChropur

reaction suitability

reagent type: derivatization reagent
reaction type: Silylations

technique(s)

gas chromatography (GC): suitable

refractive index

n20/D 1.448 (lit.)
n20/D 1.448

bp

88-90 °C/10 mmHg (lit.)

density

1.384 g/mL at 25 °C (lit.)

SMILES string

Nc1ccc2OCCOCCOc3ccccc3OCCOCCOc2c1
C[Si](C)(Cl)c1c(F)c(F)c(F)c(F)c1F

InChI

1S/C8H6ClF5Si/c1-15(2,9)8-6(13)4(11)3(10)5(12)7(8)14/h1-2H3

InChI key

PQRFRTCWNCVQHI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chlorodimethyl(pentafluorophenyl)silane is a silylated derivative employed for derivatization prior to gas chromatographic analysis of the analyte. It is basically used:
  • To increase analyte volatility.
  • To increase response.
  • To suppress the activity of an active functional group.

Other Notes

Extremely sensitive derivatizing agent for the analysis of sterols and lower aliphatic alcohols by electron capture gas chromatography[1][2][3]

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

203.0 °F - open cup

Flash Point(C)

95 °C - open cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C.F. Poole et al.
Journal of Chromatography A, 199, 123-123 (1980)
J. Heberle et al.
Silylating Agents, 2nd ed. (1995)
K. Blau et al.
Handbook of Derivatives for Chromatography (1993)

Articles

Results of a study involving the ability few Fluka silylating reagents to form GC-MS-compatible trimethylsilylmethyl derivatives of NSAIDs

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service