Skip to Content
Merck
All Photos(1)

Key Documents

76467

Supelco

Tricosafluorododecanoic acid

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

Perfluorododecanoic acid, Perfluorolauric acid

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$103.00

$103.00


Estimated to ship on31 May 2025


Request a Bulk Order

Select a Size

Change View
10 MG
$103.00

About This Item

Linear Formula:
CF3(CF2)10CO2H
CAS Number:
Molecular Weight:
614.10
Beilstein:
1811257
MDL number:
UNSPSC Code:
12352106
NACRES:
NA.21

$103.00


Estimated to ship on31 May 2025


Request a Bulk Order

grade

certified reference material
TraceCERT®

Quality Level

Agency

suitable for ASTM® 7968
suitable for ASTM® 7979
suitable for EPA 1633
suitable for EPA 533
suitable for EPA 537.1
suitable for EPA 8327
suitable for EPA ACB B21-02
suitable for EPA ACB B23-05b
suitable for EPA OTM-45
suitable for ISO 21675 2019

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

bp

245 °C/740 mmHg (lit.)

mp

105-108 °C (lit.)

application(s)

PFAS testing

SMILES string

OC(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

InChI

1S/C12HF23O2/c13-2(14,1(36)37)3(15,16)4(17,18)5(19,20)6(21,22)7(23,24)8(25,26)9(27,28)10(29,30)11(31,32)12(33,34)35/h(H,36,37)

InChI key

CXGONMQFMIYUJR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany
ASTM is a registered trademark of American Society for Testing and Materials

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Lact. - Repr. 1B - STOT RE 1

Target Organs

Liver

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Simultaneous determination of 27 PFAS chemical compounds after enriching the water samples with SPE using a Supelclean® ENVI-WAX weak anion exchange tube.

LC-MS/MS determination of per- and polyfluoroalkyl substances (PFAS) in water samples using Supelclean™ ENVI-WAX™ SPE cartridges as per EPA Method 1633.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service