Skip to Content
Merck
All Photos(1)

Key Documents

76432

Sigma-Aldrich

Pentaethylene glycol di(p-toluenesulfonate)

technical

Synonym(s):

3,6,9,12-Tetraoxatetradecane-1,14-diyl ditosylate, Pentaethylene glycol ditosylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4SO3(CH2CH2O)5SO2C6H4CH3
CAS Number:
Molecular Weight:
546.65
Beilstein:
3599245
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

technical

refractive index

n20/D 1.52
n20/D 1.524 (lit.)

density

1.26 g/mL at 25 °C (lit.)

SMILES string

Cc1ccc(cc1)S(=O)(=O)OCCOCCOCCOCCOCCOS(=O)(=O)c2ccc(C)cc2

InChI

1S/C24H34O10S2/c1-21-3-7-23(8-4-21)35(25,26)33-19-17-31-15-13-29-11-12-30-14-16-32-18-20-34-36(27,28)24-9-5-22(2)6-10-24/h3-10H,11-20H2,1-2H3

InChI key

BUHGDYPBQWWWQS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service