Skip to Content
Merck
All Photos(1)

Key Documents

75434

Sigma-Aldrich

Acetamide

≥98.0% (GC)

Synonym(s):

Amide C2

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CONH2
CAS Number:
Molecular Weight:
59.07
Beilstein:
1071207
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

vapor pressure

1 mmHg ( 65 °C)

Assay

≥98.0% (GC)

form

crystals

bp

221 °C (lit.)

mp

78-80 °C (lit.)

SMILES string

CC(N)=O

InChI

1S/C2H5NO/c1-2(3)4/h1H3,(H2,3,4)

InChI key

DLFVBJFMPXGRIB-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jie Guang et al.
Organic letters, 14(12), 3174-3177 (2012-06-02)
Highly enantioselective aldol reactions of acetylphosphonates and activated carbonyl compounds was realized with cinchona alkaloid derived catalysts, in which the acetylphosphonate was directly used as an enolate precursor for the first time. The aldol product obtained was converted in situ
Lingle Wang et al.
Proceedings of the National Academy of Sciences of the United States of America, 109(6), 1937-1942 (2012-02-07)
We apply a free energy perturbation simulation method, free energy perturbation/replica exchange with solute tempering, to two modifications of protein-ligand complexes that lead to significant conformational changes, the first in the protein and the second in the ligand. The approach
Cristian Varela et al.
Chemistry & biology, 19(4), 498-506 (2012-04-24)
Mycolic acids are vital components of the cell wall of the tubercle bacillus Mycobacterium tuberculosis and are required for viability and virulence. While mycolic acid biosynthesis is studied extensively, components involved in mycolate transport remain unidentified. We investigated the role
A E Alves et al.
Reproduction in domestic animals = Zuchthygiene, 47(6), 1003-1008 (2012-03-06)
This study was undertaken to compare cryotolerance, in terms of viability and resumption of meiosis after warming and culture (24 and 48 h), of ex situ (isolated) and in situ (enclosed in the ovarian tissue) feline cumulus-oocyte complexes (COCs) vitrified
A Subha Mahadevi et al.
Physical chemistry chemical physics : PCCP, 13(33), 15211-15220 (2011-07-16)
Insights into the formation of hydrogen bonded clusters are of outstanding importance and quantum chemical calculations play a pivotal role in achieving this understanding. Structure and energetic comparison of linear, circular and standard forms of (acetamide)(n) clusters (n = 1-15)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service