Skip to Content
Merck
All Photos(1)

Key Documents

74491

Supelco

Micro particles based on polystyrene

analytical standard, size: 20 μm

Synonym(s):

Polystyrene latex particles (20 μm), Polystyrene microbeads (20 μm), Polystyrene microspheres (20 μm), Polystyrene microparticles (20 μm), Latex beads from PS

Sign Into View Organizational & Contract Pricing

Select a Size

5 ML
$424.00
10 ML
$624.00

$424.00


Available to ship on03 April 2025Details


Request a Bulk Order

Select a Size

Change View
5 ML
$424.00
10 ML
$624.00

About This Item

MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

$424.00


Available to ship on03 April 2025Details


Request a Bulk Order

grade

analytical standard

Quality Level

form

aqueous suspension
particles

crosslinking

2 % cross-linked

concentration

10% (solids)

particle size

20 μm std dev <0.3 μm, coeff var <2%

density

1.05 g/cm3

application(s)

glass & ceramic
industrial qc
pharmaceutical

format

neat

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

This polystyrene-based microparticle (size: 20 μm) size standard is ideal for profiling particle size distribution (PSD) of test samples.

Application

Used to standardize and monitor instrument particle size.
Also used:
  • to assess the effect of microplastics on zooplankton upon ingestion[1]
  • to calibrate microfluidic device based on size measurements[2]
  • in focused beam reflectance measurement (FBRM) based particle system characterization[3]

Features and Benefits

  • suitable for routine instrument calibration checks, testing and corrections
  • available in 5 and 10 mL pack sizes as neat samples

Analysis Note

For every lot exact values of particle size and standard deviation are determined with an accuracy of 0.01 μm using a Coulter Multisizer.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Microplastics alter the properties and sinking rates of zooplankton faecal pellets
Cole M, et al.
Environmental Science & Technology, 50(6), 3239-3246 (2016)
In situ single cell detection via microfluidic magnetic bead assay
Liu F, et al.
PLoS ONE, 12(2), e0172697-e0172697 (2017)
Interpretation of focused beam reflectance measurement (FBRM) data via simulated crystallization
Yu ZQ, et al.
Organic Process Research & Development, 12(4), 646-654 (2008)

Questions

1–2 of 2 Questions  
  1. 1. What is the molecular weight of this product? 2. How many particles are there per mL?

    1 answer
    1. This product is an aqueous suspension containing 10% solids in the form of polystyrene microparticles. The molecular weight of the starting material has not been determined. The particle count is not tested or reported on a lot-to-lot basis. However, historical information reveals an estimate of 2.0 - 2.4 x 10E7 particles/mL. This information is theoretical, based on concentration and particle size.

      Helpful?

  2. We have this 5 mL of latex beads. I curious about how many beads in the bottle. It is important for our study. Thank you. Regards

    1 answer
    1. The number of beads per bottle is not determined.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service