Skip to Content
Merck
All Photos(1)

Key Documents

74024

Supelco

2,4-Di-tert-butylphenol

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)3C]2C6H3OH
CAS Number:
Molecular Weight:
206.32
Beilstein:
1910383
EC Number:
MDL number:
Product 74024 is not currently sold in your country. Contact Technical Service

grade

analytical standard

Quality Level

vapor pressure

1 mmHg ( 84.5 °C)

Assay

≥98.5% (GC)

form

crystals

shelf life

limited shelf life, expiry date on the label

bp

265 °C (lit.)

mp

53-56 °C (lit.)

SMILES string

CC(C)(C)c1ccc(O)c(c1)C(C)(C)C

InChI

1S/C14H22O/c1-13(2,3)10-7-8-12(15)11(9-10)14(4,5)6/h7-9,15H,1-6H3

InChI key

ICKWICRCANNIBI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

264.2 °F

Flash Point(C)

129 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service