Skip to Content
Merck
All Photos(1)

Documents

73970

Sigma-Aldrich

N-Methyl-N-nitroso-p-toluenesulfonamide

purum, ≥98.0% (HPLC)

Synonym(s):

p-Tolylsulfomethylnitrosamide, Diazald®, Diazomethane precursor, N-Methyl-N-(p-tolylsulfonyl)nitrosamide, N-Nitroso-p-toluenesulfomethylamide, N-Nitroso-N-methyl-p-toluenesulfonamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H10N2O3S
CAS Number:
Molecular Weight:
214.24
Beilstein:
2214345
EC Number:
MDL number:
UNSPSC Code:
12352102
PubChem Substance ID:
NACRES:
NA.22

grade

purum

Quality Level

Assay

≥98.0% (HPLC)

form

crystals

reaction suitability

reaction type: C-C Bond Formation

mp

~58 °C

storage temp.

2-8°C

SMILES string

CN(N=O)S(=O)(=O)c1ccc(C)cc1

InChI

1S/C8H10N2O3S/c1-7-3-5-8(6-4-7)14(12,13)10(2)9-11/h3-6H,1-2H3

InChI key

FFKZOUIEAHOBHW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage and Stability

Warning: these products are subject to the Explosives Act and must be transported refrigerated - additional costs for transport will apply.

Other Notes

Preparation of diazomethane

Legal Information

Diazald is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Self-react. C - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

4.1A - Other explosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Th.J. De Boer et al.
Organic Syntheses, 4, 250-250 (1963)
M. Hudlicky
The Journal of Organic Chemistry, 45, 5377-5377 (1980)
Izabela Kania-Korwel et al.
Journal of chromatography. A, 1207(1-2), 146-154 (2008-09-02)
Several polychlorinated biphenyls (PCBs) and their hydroxylated metabolites display axial chirality. Here we describe an enantioselective, gas chromatographic separation of methylated derivatives of hydroxylated (OH-)PCB atropisomers (MeO-PCB) using a chemically bonded beta-cyclodextrin column (Chirasil-Dex). The atropisomers of several MeO-PCBs could
Contact dermatitis from Diazald.
H S Young et al.
Contact dermatitis, 50(5), 315-316 (2004-06-24)
J V Uri et al.
Acta microbiologica Hungarica, 39(3-4), 317-322 (1992-01-01)
Diazald, a chemical intermediate for the synthesis of biologically active compounds, was found to be a potent in vitro antimicrobial agent against yeasts, yeast-like and filamentous fungi as well as Gram-positive and Gram-negative bacterial strains. Its activity is not inhibited

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service